missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Normal Mouse IgG Mouse, Polyclonal, Isotype Control, MilliporeSigma™
Description
Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation and Western Blotting.
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide containing 30% glycerol. Liquid at −20°C. |
| Host Species | Mouse |
| Quantity | 250 μg |
| Primary or Secondary | Primary |
| Content And Storage | Stable for 1 year at -20°C from date of receipt. For maximum recovery of product centrifuge the original vial prior to removing the cap. Handling Recommendations: Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution. |
| Show More |
For Research Use Only