Learn More
Abcam Inc Normal Donkey Serum, 10ML
Catalog No. NC2039255
Encompass
Normal donkey serum is used extensively for the blocking of non-specific antibody binding in tissue and cell staining, and in other applications of antibodies. - Cited in over 190 publications - Suitable for immunocytochemistry/immunofluorescence (ICC/IF), immunohistochemistry (IHC), and more - Superior Fc receptor blocking - Reduces background noise
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2039255 Supplier Abcam Inc Supplier No. AB747510ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Normal donkey serum is used extensively for the blocking of non-specific antibody binding in tissue and cell staining, and in other applications of antibodies. - Cited in over 190 publications - Suitable for immunocytochemistry/immunofluorescence (ICC/IF), immunohistochemistry (IHC), and more - Superior Fc receptor blocking - Reduces background noise
The product is subject to the following: Abcam Restricted Use Statement