missing translation for 'onlineSavingsMsg'
Learn More

Norit™ Activated Charcoal, RBAA-3, MilliporeSigma™ Supelco™

Catalog No. 111015409
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015409 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015409 Supplier Merck Emd Millipore Supplier No. 292381KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00133992
Synonym Charcoal activated Norit(R)

Specifications

CAS 7440-44-0
Linear Formula C
Quantity 1 kg
Formula Weight 12.01
Physical Form Rod
Product Line Norit

Norit is a registered trademark of Norit N.V.