missing translation for 'onlineSavingsMsg'
Learn More

Norharman, 98%

Catalog No. AC129265000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129265000 500 mg Glass bottle
AC129261000 100 mg Glass bottle
AC129260025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC129265000 Supplier Thermo Scientific Chemicals Supplier No. 129265000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 244-63-3 | C11H8N2 | 168.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 244-63-3
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00004956
InChI Key AIFRHYZBTHREPW-UHFFFAOYSA-N
Synonym 9h-pyrido 3,4-b indole, norharman, norharmane, beta-carboline, carbazoline, 2,9-diazafluorene, 9h-beta-carboline, 2-azacarbazole, .beta.-carboline
PubChem CID 64961
ChEBI CHEBI:109895
IUPAC Name 9H-pyrido[3,4-b]indole
SMILES N1C2=C(C=CC=C2)C2=C1C=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 199.0°C to 201.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Tertiary base)
Packaging Glass bottle
Quantity 500 mg
Beilstein 23, II, 224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 168.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Norharman
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-959-

RUO – Research Use Only