missing translation for 'onlineSavingsMsg'
Learn More

Norgestimate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023009
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023009 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023009 Supplier Merck Emd Millipore Supplier No. PHR2119500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 35189-28-7
Flash Point Not Applicable
Molecular Formula C23H31NO3
Linear Formula C23H31NO3
MDL Number MFCD00867874
Quantity 500 mg
Synonym Norgestimate; (17 alpha)-17-(Acetyloxy)-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one 3-oxime; Dexnorgestrel acetime
Molecular Weight (g/mol) 369.50
Formula Weight 369.50
Grade Certified Reference Material
Physical Form Solid
Show More Show Less