missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Norfloxacin | |
| 221°C | |
| C16H18FN3O3 | |
| 25 g | |
| OGJPXUAPXNRGGI-UHFFFAOYSA-N | |
| 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid | |
| 4539 | |
| 319.34 | |
| Crystalline Powder |
| 70458-96-7 | |
| Yellow | |
| MFCD00079532 | |
| norfloxacin, norfloxacine, noroxin, chibroxin, baccidal, sebercim, norfloxacinum, norfloxacino, barazan, fulgram | |
| CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O | |
| 319.336 | |
| CHEBI:100246 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 70458-96-7 | |
| 319.336 | |
| OGJPXUAPXNRGGI-UHFFFAOYSA-N | |
| 4539 | |
| 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid |
| C16H18FN3O3 | |
| MFCD00079532 | |
| norfloxacin, norfloxacine, noroxin, chibroxin, baccidal, sebercim, norfloxacinum, norfloxacino, barazan, fulgram | |
| CHEBI:100246 | |
| CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O |
Safety and Handling
RTECSNumber : VB2005000
TSCA : No