missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin 98.0+%, TCI America™

Catalog No. N08175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08175G 5 g
N081725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08175G Supplier TCI America Supplier No. N08175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16H18FN3O3
Molecular Weight (g/mol) 319.336
MDL Number MFCD00079532
InChI Key OGJPXUAPXNRGGI-UHFFFAOYSA-N
Synonym norfloxacin, norfloxacine, noroxin, chibroxin, baccidal, sebercim, norfloxacinum, norfloxacino, barazan, fulgram
PubChem CID 4539
ChEBI CHEBI:100246
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

Melting Point 221°C
Color Yellow
Quantity 5 g
Formula Weight 319.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Norfloxacin
RTECSNumber VB2005000
TSCA No