missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals

Catalog No. AC445260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445260050 Supplier Thermo Scientific Chemicals Supplier No. 445260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16H18FN3O3
Molecular Weight (g/mol) 319.336
InChI Key OGJPXUAPXNRGGI-UHFFFAOYSA-N
Synonym 1-Ethyl-6-fluoro-1, 4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid
PubChem CID 4539
ChEBI CHEBI:100246
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

Type Norfloxacin
Physical Form Crystalline Powder
Melting Point 218°C to 226°C
Ignition Residue 0.2 % max.
Infrared Spectrum Authentic
Water 3% Max. (K.F.)
Color White to Yellow
Quantity 5 g
Assay Percent Range 98%
Formula Weight 319.34
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Norfloxacin
Show More Show Less

RUO – Research Use Only