missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norcantharidin 98.0+%, TCI America™
Supplier: TCI America N09141G
Specifications
| Norcantharidin | |
| 115°C | |
| C8H8O4 | |
| 1 g | |
| JAABVEXCGCXWRR-GUCUJZIJSA-N | |
| 168.148 | |
| 168.15 | |
| Crystalline Powder |
| 29745-04-8 | |
| White | |
| MFCD00213361 | |
| exo-3,6-Epoxyhexahydrophthalic Anhydride, exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic Anhydride | |
| C1CC2C3C(C1O2)C(=O)OC3=O | |
| 12251906 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 29745-04-8 | |
| 168.148 | |
| JAABVEXCGCXWRR-GUCUJZIJSA-N | |
| 12251906 |
| C8H8O4 | |
| MFCD00213361 | |
| exo-3,6-Epoxyhexahydrophthalic Anhydride, exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic Anhydride | |
| C1CC2C3C(C1O2)C(=O)OC3=O |
Safety and Handling
EINECSNumber : (3)-2460
RTECSNumber : RN8400000
TSCA : No