missing translation for 'onlineSavingsMsg'
Learn More

Norcamphor, 99%

CAS: 497-38-1 | C7H10O | 110.16 g/mol

Chemical Identifiers

CAS 497-38-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00074823
InChI Key KPMKEVXVVHNIEY-UHFFFAOYSA-N
Synonym norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
PubChem CID 10345
IUPAC Name bicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1CC2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC129250050
View Documents Promotion Details
N/A 5 g Glass bottle N/A
Only null left
 
AC129250250
View Documents Promotion Details
N/A 25 g Plastic bottle N/A
Only null left
 
AC129251000
View Documents Promotion Details
N/A 100 g Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

497-38-1
110.16
KPMKEVXVVHNIEY-UHFFFAOYSA-N
10345
C1CC2CC1CC2=O
C7H10O
MFCD00074823
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
bicyclo[2.2.1]heptan-3-one

Specifications

497-38-1
100.0
Colorless to White
168°C to 172°C
Authentic
C7H10O
07, 57
Solubility in water: insoluble. Other solubilities: soluble in methanol
C1CC2CC1CC2=O
110.16
110.16
Adhering Crystals
98.5
88°C to 91°C
33g/mL
33°C
99%
MFCD00074823
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
KPMKEVXVVHNIEY-UHFFFAOYSA-N
bicyclo[2.2.1]heptan-3-one
10345
99%
Norcamphor
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 207-846-1

SDS
Documents

Documents

RUO – Research Use Only