missing translation for 'onlineSavingsMsg'
Learn More

Norcamphor, 98+%

Used as a building block in organic synthesis and also acts as a precursor to norborneols. | CAS: 497-38-1 | C7H10O | 110.156 g/mol

Supplier:  Thermo Scientific Chemicals L0149106

 View more versions of this product

Catalog No. AAL0149106


Only null left
Explore available promotions

Description

Description

Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
Specifications

Specifications

Norcamphor
92°C to 97°C
33°C (91°F)
MFCD00074823
UN1325
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
KPMKEVXVVHNIEY-UHFFFAOYSA-N
bicyclo[2.2.1]heptan-3-one
10345
≥98%
497-38-1
172°C to 174°C
C7H10O
5 g
1209657
Insoluble in water. Soluble in methanol.
C1CC2CC1CC2=O
110.156
110.16

Chemical Identifiers

497-38-1
110.156
KPMKEVXVVHNIEY-UHFFFAOYSA-N
10345
C1CC2CC1CC2=O
C7H10O
MFCD00074823
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
bicyclo[2.2.1]heptan-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 207-846-1

RTECSNumber : RB7680000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only