missing translation for 'onlineSavingsMsg'
Learn More

Norcamphor, 98+%

Catalog No. AAL0149114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0149114 25 g
AAL0149106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0149114 Supplier Thermo Scientific Chemicals Supplier No. L0149114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a building block in organic synthesis and also acts as a precursor to norborneols. | CAS: 497-38-1 | C7H10O | 110.156 g/mol

Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.

Chemical Identifiers

CAS 497-38-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00074823
InChI Key KPMKEVXVVHNIEY-UHFFFAOYSA-N
Synonym norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
PubChem CID 10345
IUPAC Name bicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1CC2=O

Specifications

Melting Point 92°C to 97°C
Boiling Point 172°C to 174°C
Flash Point 33°C (91°F)
Quantity 25 g
UN Number UN1325
Beilstein 1209657
Solubility Information Insoluble in water. Soluble in methanol.
Formula Weight 110.16
Percent Purity ≥98%
Chemical Name or Material Norcamphor
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 207-846-1
RTECSNumber RB7680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only