missing translation for 'onlineSavingsMsg'
Learn More

Norbornene, 99%

CAS: 498-66-8 | C7H10 | 94.157 g/mol

$74.14 - $231.26

Chemical Identifiers

CAS 498-66-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.157
MDL Number MFCD00082304
InChI Key JFNLZVQOOSMTJK-UHFFFAOYSA-N
Synonym bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
PubChem CID 10352
IUPAC Name bicyclo[2.2.1]hept-2-ene
SMILES C1CC2CC1C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1110322
View Documents
Thermo Scientific Chemicals
L1110322
100 g
Each for $74.14
Only null left
 
AAL1110336
View Documents
Thermo Scientific Chemicals
L1110336
500 g
Each for $231.26
Only null left
 
Description

Description

Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs' catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs′ catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

Solubility
Soluble in ethanol, ether and most organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

498-66-8
94.157
JFNLZVQOOSMTJK-UHFFFAOYSA-N
10352
C1CC2CC1C=C2
C7H10
MFCD00082304
bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
bicyclo[2.2.1]hept-2-ene

Specifications

498-66-8
0.955
−15°C (5°F)
C7H10
100 g
1098544
Soluble in ethanol,ether and most organic solvents. Insoluble in water.
C1CC2CC1C=C2
94.157
94.16
Norbornene
44°C to 47°C
95°C to 96°C
Pungent
MFCD00082304
UN1325
bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
JFNLZVQOOSMTJK-UHFFFAOYSA-N
bicyclo[2.2.1]hept-2-ene
10352
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319
Flammable solid.
Causes serious eye irritation.

P201-P202-P210-P240-P241-P261-P264b-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P331-P370+P378q-P501c

H228-H304-H319-H336-H361

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 207-866-0

RTECSNumber : RB7900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only