missing translation for 'onlineSavingsMsg'
Learn More

Norbornene, 99%

Catalog No. AAL1110336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1110336 500 g
AAL1110322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1110336 Supplier Thermo Scientific Chemicals Supplier No. L1110336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 498-66-8 | C7H10 | 94.157 g/mol

Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs' catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs′ catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

Solubility
Soluble in ethanol, ether and most organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 498-66-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.157
MDL Number MFCD00082304
InChI Key JFNLZVQOOSMTJK-UHFFFAOYSA-N
Synonym bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
PubChem CID 10352
IUPAC Name bicyclo[2.2.1]hept-2-ene
SMILES C1CC2CC1C=C2

Specifications

Melting Point 44°C to 47°C
Density 0.955
Boiling Point 95°C to 96°C
Flash Point −15°C (5°F)
Odor Pungent
Quantity 500 g
UN Number UN1325
Beilstein 1098544
Solubility Information Soluble in ethanol,ether and most organic solvents. Insoluble in water.
Formula Weight 94.16
Percent Purity 99%
Chemical Name or Material Norbornene
Show More Show Less
Hazard Category H228-H304-H319-H336-H361
Hazard Statement GHS H Statement
H228-H319
Flammable solid.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P240-P241-P261-P264b-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 207-866-0
RTECSNumber RB7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only