missing translation for 'onlineSavingsMsg'
Learn More

Norbornane-2-carboxylic acid, 98%, predominantly endo isomer

Catalog No. AAL0843914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0843914 25 g
AAL0843906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0843914 Supplier Thermo Scientific Chemicals Supplier No. L0843914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 824-62-4 | C8H12O2 | 140.18 g/mol

Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Incompatible with oxidizing agents, Strong acids, Strong bases.

Chemical Identifiers

CAS 824-62-4
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00167749
InChI Key JESWDXIHOJGWBP-UHFFFAOYNA-N
Synonym bicyclo 2.2.1 heptane-2-carboxylic acid, 2-norbornanecarboxylic acid, norbornane-2-carboxylic acid, 2-norbornanecarboxylic acid, endo, bicyclo 2.2.1 heptane-2-carboxylic acid, endo, exo-norbornane-2-carboxylic acid, bicyclo 2.2.1 heptane-3-carboxylic acid, 2-norbornanecarboxlic acid
PubChem CID 79113
SMILES OC(=O)C1CC2CCC1C2

Specifications

Melting Point ∼40°C
Boiling Point 134°C to 136°C (16 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.4855
Quantity 25 g
UN Number UN3261
Beilstein 2042631
Solubility Information Insoluble in water.
IUPAC Name bicyclo[2.2.1]heptane-3-carboxylic acid
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material Norbornane-2-carboxylic acid, predominantly endo isomer
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-532-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only