missing translation for 'onlineSavingsMsg'
Learn More

Nor-S-(-)-SCH-23388 hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111012405
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012405 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012405 Supplier Merck Emd Millipore Supplier No. S1155MG
Only null left

Specifications

CAS 107128-79-0
Physical Form Neat
Linear Formula C16H16 ClNO · HCl
MDL Number MFCD00069369
Synonym -(-)-7-Chloro-8-hydroxy-1-phenyl-2,3,4,5-tetrahydro-1 H-3-benzazepine
Type Analytical Standard
Quantity 5 mg
Formula Weight 310.22
Grade Analytical Standard