missing translation for 'onlineSavingsMsg'
Learn More

(+)-Nootkatone, crystalline, 98+%

Catalog No. aaa1916603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1916603 1 g
AAA1916606 5 g
AAA1916614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1916603 Supplier Thermo Scientific Chemicals Supplier No. A1916603
Only null left

CAS: 4674-50-4 | C15H22O | 218.34 g/mol

(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

Solubility
Slightly soluble in ethanol and chloroform. Partly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4674-50-4
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00036591
InChI Key WTOYNNBCKUYIKC-SLEUVZQESA-N
Synonym unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one
PubChem CID 7567181
IUPAC Name (4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one
SMILES CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C

Specifications

Melting Point 34°C to 36°C
Density 0.997
Boiling Point 125°C (0.5 mmHg)
Flash Point 99°C (210°F)
Quantity 1 g
Beilstein 4676969
Solubility Information Slightly soluble in ethanol and chloroform. Partly soluble in water.
Optical Rotation +185° (c=1 in Chloroform)
Formula Weight 218.34
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material (+)-Nootkatone
Show More Show Less
EINECSNumber 225-124-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only