missing translation for 'onlineSavingsMsg'
Learn More

(+)-Nootkatone, crystalline, 98+%

CAS: 4674-50-4 | C15H22O | 218.34 g/mol

Supplier:  Thermo Scientific Chemicals A1916606

Quantity 5 g

 View more versions of this product

Catalog No. AAA1916606


Only null left

Description

Description

(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

Solubility
Slightly soluble in ethanol and chloroform. Partly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

4674-50-4
218.34
WTOYNNBCKUYIKC-SLEUVZQESA-N
7567181
CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C
C15H22O
MFCD00036591
unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one
(4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one

Specifications

(+)-Nootkatone
34°C to 36°C
125°C (0.5 mmHg)
C15H22O
MFCD00036591
unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one
WTOYNNBCKUYIKC-SLEUVZQESA-N
CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C
218.34
218.34
Crystalline
4674-50-4
0.997
99°C (210°F)
5 g
4676969
Slightly soluble in ethanol and chloroform. Partly soluble in water.
+185° (c=1 in Chloroform)
(4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one
7567181
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 225-124-4

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions