missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Nonylphenol

Catalog No. 2908583L
Encompass
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2908583L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2908583L Supplier Sigma Aldrich Supplier No. 2908583L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Nonylphenol is a toxic xenobiotic compound classified as an endocrine disrupter capable of interfering with the hormonal system of numerous organisms
  • Nonylphenol has been reported as antiandrogen by using a yeast detection system
  • Molecular mechanism for the antiandrogenic action of nonylphenol has been reported

Specifications

CAS 84852-15-3
Density 0.937 g/mL (at 25°C (literature))
Molecular Formula C15H24O
Refractive Index n20/D 1.511 (literature)
Linear Formula C15H24O
Quantity 3 L
Molecular Weight (g/mol) 220.35