missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonylbenzene 97.0+%, TCI America™
Supplier: TCI America N029825ML
Specifications
| Nonylbenzene | |
| Yellow | |
| C15H24 | |
| 25 mL | |
| LIXVMPBOGDCSRM-UHFFFAOYSA-N | |
| nonylbenzene | |
| 14126 | |
| ≥97.0% (GC) |
| 1081-77-2 | |
| 282°C | |
| MFCD00009575 | |
| 1-phenylnonane, benzene, nonyl, n-nonylbenzene, nonane, 1-phenyl, 1-nonylbenzene, unii-58ak68ov26, benzene, c6-12-alkyl derivs., n-nonyl benzene, phenyl-nonane, c6-c12 alkylbenzene | |
| CCCCCCCCCC1=CC=CC=C1 | |
| 204.357 | |
| 204.36 | |
| Liquid |
Chemical Identifiers
| 1081-77-2 | |
| 204.357 | |
| LIXVMPBOGDCSRM-UHFFFAOYSA-N | |
| 14126 | |
| CCCCCCCCCC1=CC=CC=C1 |
| C15H24 | |
| MFCD00009575 | |
| 1-phenylnonane, benzene, nonyl, n-nonylbenzene, nonane, 1-phenyl, 1-nonylbenzene, unii-58ak68ov26, benzene, c6-12-alkyl derivs., n-nonyl benzene, phenyl-nonane, c6-c12 alkylbenzene | |
| nonylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes