missing translation for 'onlineSavingsMsg'
Learn More

Nonylamine 98.0+%, TCI America™

Catalog No. p-7133739
Change view
Click to view available options
Quantity:
5 mL
25 mL
Catalog No. Quantity
N02975ML 5 mL
N029725ML 25 mL
2 options

Catalog No. N02975ML

Supplier: TCI America N02975ML

Only null left

Chemical Identifiers

CAS 112-20-9
Molecular Formula C9H22ClN
Molecular Weight (g/mol) 179.73
MDL Number MFCD00008249
InChI Key IMGHCWMUWVHYCO-UHFFFAOYSA-N
Synonym nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9
PubChem CID 16215
IUPAC Name nonan-1-aminium chloride
SMILES [Cl-].CCCCCCCCC[NH3+]

Specifications

Color Yellow
Boiling Point 201°C
Quantity 5 mL
UN Number 2735
Formula Weight 143.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonylamine
EINECSNumber (2)-0133
TSCA Yes