Learn More
Nonyl aldehyde, 95%, Thermo Scientific Chemicals
$102.78 - $102.78
Chemical Identifiers
| CAS | 124-19-6 |
|---|---|
| Molecular Formula | C9H18O |
| Molecular Weight (g/mol) | 142.24 |
| MDL Number | MFCD00007030 |
| InChI Key | GYHFUZHODSMOHU-UHFFFAOYSA-N |
| Synonym | pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde |
| PubChem CID | 31289 |
| ChEBI | CHEBI:84268 |
| IUPAC Name | nonanal |
| SMILES | CCCCCCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC357571000
|
Thermo Scientific Chemicals
357571000 |
100 mL | Glass bottle |
Each for $102.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 124-19-6 | |
| 142.24 | |
| GYHFUZHODSMOHU-UHFFFAOYSA-N | |
| 31289 | |
| nonanal |
| C9H18O | |
| MFCD00007030 | |
| pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde | |
| CHEBI:84268 | |
| CCCCCCCCC=O |
Specifications
| 124-19-6 | |
| 100.0 | |
| 0.8270g/mL | |
| 63°C | |
| 94% min. (GC) | |
| C9H18O | |
| CH3(CH2)7CHO | |
| 100 mL | |
| 0.827 | |
| Solubility in water: practically insoluble. Other solubilities: soluble in alcohol | |
| CCCCCCCCC=O | |
| 142.24 | |
| CHEBI:84268 | |
| 95% | |
| Nonyl aldehyde, 95% |
| 94.0 | |
| Colorless to Yellow | |
| 93.0°C (23.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4230 to 1.4270 | |
| MFCD00007030 | |
| 01,708 | |
| pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde | |
| GYHFUZHODSMOHU-UHFFFAOYSA-N | |
| nonanal | |
| 31289 | |
| 142.24 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 204-688-5
RTECSNumber : RA5700000
TSCA : TSCA
RUO – Research Use Only