Learn More
Invitrogen™ RNase-free Microfuge Tubes
These microcentrifuge tubes are guaranteed RNase- and DNase-free.
Supplier:  Invitrogen™ AM12475
Encompass
 View more versions of this product
Catalog No. AM12475
Only null left
Description
Description
Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.
Features include:
  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
Specifications
Chemical Identifiers
Clear
Store at room temperature.
Room Temperature
Autoclavable
Yes
Polypropylene
Invitrogen
RNase-Free, DNase-Free
Snap Cap
Microfuge Tube (RNase-free)
Specifications
Clear
Polypropylene
Store at room temperature.
Room Temperature
Autoclavable
Yes
Microfuge Tube (RNase-free)
2 mL
250 Tubes/Pk.
Invitrogen
RNase-Free, DNase-Free
Snap Cap
Yes
Graduated, low-binding
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.