missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parter Medical Products™ Nonsterile Specimen Containers
Description
- Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
- Stackable, leak-resistant and graduated unless otherwise noted
- Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
- Manufactured under strict process control and lot numbers are fully traceable
- 95kpa compliant; testing results available upon request
- Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
- Available in individually wrapped (Flo-Pac™) formats
- Closures secure at 10 to 12 in.-lb. closing force
- Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
- Standard cap colors are natural, orange and blue
Specifications
Specifications
| Graduated | Yes |
| Material | Polypropylene (Clarified) |
| Sterility | Non-sterile |
| Capacity (English) | 4 oz. |
| Capacity (Metric) | 120 mL |
| Closure Color | White |
| Closure Material | Polyethylene |
| Closure Size | 53 mm |
| Label | Tabbed |