missing translation for 'onlineSavingsMsg'
Learn More

Parter Medical Products™ Nonsterile Specimen Containers

Catalog No. 22150113
Change view
Click to view available options
Closure Color:
White
Label:
Tabbed
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Label
22150113 White Tabbed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22150113 Supplier Parter Medical Products™ Supplier No. 142211
Only null left

Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system

  • Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
  • Stackable, leak-resistant and graduated unless otherwise noted
  • Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
  • Manufactured under strict process control and lot numbers are fully traceable
  • 95kpa compliant; testing results available upon request
  • Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
  • Available in individually wrapped (Flo-Pac™) formats
  • Closures secure at 10 to 12 in.-lb. closing force
  • Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
  • Standard cap colors are natural, orange and blue

Specifications

Graduated Yes
Material Polypropylene (Clarified)
Sterility Non-sterile
Capacity (English) 4 oz.
Capacity (Metric) 120 mL
Closure Color White
Closure Material Polyethylene
Closure Size 53 mm
Label Tabbed