missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems

Catalog No. 21402524
Change view
Click to view available options
Volume (Metric):
20 μL
250 μL
For Use With (Equipment):
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Fit for Purpose:
Certified Quality
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment) Fit for Purpose
21402524 250 μL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 Certified Quality
21402521 20 μL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21402524 Supplier Thermo Scientific™ Supplier No. 919252

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.

Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:

  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Format Rack
Tip Style Conductive
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Product Line Beckman
Fit for Purpose Certified Quality