missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Nonmetallic Liquid Disposal Safety Cans

Resists denting, puncturing, and chipping. The nonmetallic liquid disposal safety can has large mouth opening and lock-open cap for easy filling/pouring.

Supplier:  Justrite™ 12754

 View more versions of this product

Catalog No. 19159312


Only null left

Description

Description

Resists denting, puncturing, and chipping. The nonmetallic liquid disposal safety can has large mouth opening and lock-open cap for easy filling/pouring.

  • Resist dents, punctures, and chemicals
  • Safely store liquids that corrode stainless- or coated-steel safety cans
  • Available with stainless-steel fittings
  • Double-wall flame arrester guards against outside ignition
  • Cap automatically vents to prevent pressure buildup between 3 and 5psig

Certifications

FM approved (except 17-985-72C).

Specifications

Chemical Identifiers

FM
30.5 cm
12 in.
Liquid Disposal Can

Specifications

19 L
30.5 cm
5 gal.
50.8 cm
9.3 cm
For easy filling/pouring
HDPE, Stainless Steel Fitting
FM
12 in.
White
3.65 in.
Flame arrester, stainless steel fitting
20 in.
Liquid Disposal Can
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions