missing translation for 'onlineSavingsMsg'
Learn More

Drierite™ Drying Desiccants, W.A. Hammond

Catalog No. p-3143283
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
Mesh Size:
8 Mesh
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Mesh Size
075773A 1 lb. 8 Mesh
075773B 5 lb. 8 Mesh
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 075773A Supplier W A Hammond Drierite Co Supplier No. HDC13001
Only null left

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

  • All purpose desiccant made of anhydrous calcium sulfate
  • Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
  • Dries organic liquids to low ppm levels
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas; dryers, glove boxes, sealed chambers
  • Useful for water removal from liquid reaction mixtures and multi-component;gas streams
  • Neutral, stable, constant in volume, and inert except toward water
  • Insoluble in organic solvents
  • Absorbs moisture at temperatures up to 66°C

Chemical Identifiers

Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

CAS 7778-18-9
Material Calcium Sulfate
Quantity 1 lb.
Mesh Size 8 Mesh
Packaging Glass Bottle
Synonym Abosrbent
Chemical Name or Material Calcium Sulfate