missing translation for 'onlineSavingsMsg'
Learn More

Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals

Catalog No. AAJ19628AP
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ19628AP 500 mL Plastic bottle
AAJ19628K2 1 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ19628AP Supplier Thermo Fisher Scientific Supplier No. J19628AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-93-1 | Molecular Formula: C16H26O2 | Molecular Weight: 250.38 g/mol

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Physical Form Viscous Liquid
Boiling Point >250°C
Melting Point ∼5°C
Density 1.061
Quantity 500 mL
Flash Point >110°C
Packaging Plastic bottle
Specific Gravity 1.061
Formula Weight ca 625
Chemical Name or Material Nonidet™ P40 Substitute
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
RTECSNumber MD0907600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only