missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonaqueous Buffer Stock Solution A, for Acid Number of Petroleum Products, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Nonaqueous Buffer Stock Solution A |
| Name Note | For Acid Number of Petroleum Products |
| CAS Min % | 93.0, 2.9, 1.76 |
| CAS Max % | 97.0, 3.01, 1.83 |
| CAS | 108-75-8, 7647-01-0 |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |