missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanoyl Chloride 95.0+%, TCI America™
$57.79 - $169.63
Chemical Identifiers
| CAS | 764-85-2 |
|---|---|
| Molecular Formula | C9H17ClO |
| Molecular Weight (g/mol) | 176.684 |
| MDL Number | MFCD00000768 |
| InChI Key | NTQYXUJLILNTFH-UHFFFAOYSA-N |
| Synonym | nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride |
| PubChem CID | 69819 |
| IUPAC Name | nonanoyl chloride |
| SMILES | CCCCCCCCC(=O)Cl |
Chemical Identifiers
| 764-85-2 | |
| 176.684 | |
| NTQYXUJLILNTFH-UHFFFAOYSA-N | |
| 69819 | |
| CCCCCCCCC(=O)Cl |
| C9H17ClO | |
| MFCD00000768 | |
| nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride | |
| nonanoyl chloride |
Specifications
| 764-85-2 | |
| 215°C | |
| MFCD00000768 | |
| 3265 | |
| NTQYXUJLILNTFH-UHFFFAOYSA-N | |
| nonanoyl chloride | |
| 69819 | |
| ≥95.0% (T) | |
| Nonanoyl Chloride |
| Yellow | |
| C9H17ClO | |
| 25 g | |
| nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride | |
| CCCCCCCCC(=O)Cl | |
| 176.684 | |
| 176.68 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes