missing translation for 'onlineSavingsMsg'
Learn More

Nonanoyl Chloride 97.0+%, TCI America™

Catalog No. N081325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N081325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N081325G Supplier TCI America Supplier No. N081325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 764-85-2
Molecular Formula C9H17ClO
Molecular Weight (g/mol) 176.684
MDL Number MFCD00000768
InChI Key NTQYXUJLILNTFH-UHFFFAOYSA-N
Synonym nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
PubChem CID 69819
IUPAC Name nonanoyl chloride
SMILES CCCCCCCCC(=O)Cl

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 215°C
Quantity 25 g
UN Number 3265
Formula Weight 176.68
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Nonanoyl Chloride
EINECSNumber (2)-0632
TSCA Yes