missing translation for 'onlineSavingsMsg'
Learn More

Nonanoyl chloride, 96%, Thermo Scientific Chemicals

$83.14 - $204.78

Chemical Identifiers

CAS 764-85-2
Molecular Formula C9H17ClO
Molecular Weight (g/mol) 176.684
MDL Number MFCD00000768
InChI Key NTQYXUJLILNTFH-UHFFFAOYSA-N
Synonym nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
PubChem CID 69819
IUPAC Name nonanoyl chloride
SMILES CCCCCCCCC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1767114
View Documents
Thermo Scientific Chemicals
A1767114
25 g
Each for $83.14
Only null left
 
AAA1767122
View Documents
Thermo Scientific Chemicals
A1767122
100 g
Each for $204.78
Only null left
 
Description

Description

Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

764-85-2
176.684
NTQYXUJLILNTFH-UHFFFAOYSA-N
69819
CCCCCCCCC(=O)Cl
C9H17ClO
MFCD00000768
nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
nonanoyl chloride

Specifications

764-85-2
0.94
95°C (203°F)
1.438
25 g
1362621
nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
NTQYXUJLILNTFH-UHFFFAOYSA-N
nonanoyl chloride
69819
97%
-61°C
215°C
C9H17ClO
MFCD00000768
UN3265
Moisture sensitive
Reacts with water.
CCCCCCCCC(=O)Cl
176.684
176.69
Nonanoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-131-2

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only