missing translation for 'onlineSavingsMsg'
Learn More

Nonanoyl chloride, 96%, Thermo Scientific Chemicals

Catalog No. AAA1767114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1767114 25 g
AAA1767122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1767114 Supplier Thermo Scientific Chemicals Supplier No. A1767114

Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 764-85-2
Molecular Formula C9H17ClO
Molecular Weight (g/mol) 176.684
MDL Number MFCD00000768
InChI Key NTQYXUJLILNTFH-UHFFFAOYSA-N
Synonym nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride
PubChem CID 69819
IUPAC Name nonanoyl chloride
SMILES CCCCCCCCC(=O)Cl

Specifications

Melting Point -61°C
Density 0.94
Boiling Point 215°C
Flash Point 95°C (203°F)
Refractive Index 1.438
Quantity 25 g
UN Number UN3265
Beilstein 1362621
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 176.69
Percent Purity 97%
Chemical Name or Material Nonanoyl chloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-131-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only