missing translation for 'onlineSavingsMsg'
Learn More

Nonanophenone, 97%

Catalog No. AAA1223818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1223818 50 g
AAA1223809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1223818 Supplier Thermo Scientific Chemicals Supplier No. A1223818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6008-36-2 | C15H22O | 218.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6008-36-2
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00048965
InChI Key PFUPABFCHVRLLY-UHFFFAOYSA-N
Synonym nonanophenone, n-nonanophenone, n-octyl phenyl ketone, 1-phenyl-1-nonanone, 1-nonanone, 1-phenyl, 1-phenyl-nonan-1-one, acmc-209mhy
PubChem CID 80108
IUPAC Name 1-phenylnonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 14°C
Density 0.94
Boiling Point 139°C to 140°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.506
Quantity 50 g
Beilstein 2047454
Formula Weight 218.34
Percent Purity 97%
Chemical Name or Material Nonanophenone
EINECSNumber 227-861-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only