missing translation for 'onlineSavingsMsg'
Learn More

Nonanoic Anhydride 97.0+%, TCI America™

Catalog No. p-7134873
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P095725ML 25 mL
P0957500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P095725ML Supplier TCI America Supplier No. P095725ML

Chemical Identifiers

CAS 1680-36-0
Molecular Formula C18H34O3
Molecular Weight (g/mol) 298.47
MDL Number MFCD00057768
InChI Key PBBAFLCORNAZCD-UHFFFAOYSA-N
Synonym nonanoic anhydride, pelargonic anhydride, nonan-1-oic anhydride, nonanoic acid,1,1'-anhydride, acmc-209dxl
PubChem CID 74304
IUPAC Name nonanoyl nonanoate
SMILES CCCCCCCCC(=O)OC(=O)CCCCCCCC

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 mL
UN Number 3265
Formula Weight 298.47
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Nonanoic Anhydride