missing translation for 'onlineSavingsMsg'
Learn More

Nonanenitrile 98.0+%, TCI America™

Catalog No. p002025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P002025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P002025G Supplier TCI America Supplier No. P002025G
Only null left

Specifications

CAS 2243-27-8
Color Yellow
Boiling Point 222°C
Molecular Formula C9H17N
Quantity 25 g
MDL Number MFCD00001985
Synonym n-octyl cyanide, n-octylcyanide, octyl cyanide, pelargononitrile, n-nonanenitrile, n-nonanonitrile, nonanonitrile, nonanitrile, pelargonitrile, 1-cyanooctane
InChI Key PLZZPPHAMDJOSR-UHFFFAOYSA-N
SMILES CCCCCCCCC#N
IUPAC Name nonanenitrile
Molecular Weight (g/mol) 139.242
PubChem CID 16715
Formula Weight 139.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanenitrile
Show More Show Less
EINECSNumber (2)-1507
RTECSNumber RA6606000
TSCA Yes