missing translation for 'onlineSavingsMsg'
Learn More

Nonane 98.0+%, TCI America™

Catalog No. p-7133731
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N028625ML 25 mL
N0286100ML 100 mL
N0286500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N028625ML Supplier TCI America Supplier No. N028625ML
Only null left

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -51°C
Color Colorless
Boiling Point 150°C
Quantity 25 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonane
EINECSNumber (2)-0009
RTECSNumber RA6115000
TSCA Yes