missing translation for 'onlineSavingsMsg'
Learn More

Nonane 98.0+%, TCI America™

Catalog No. n028625ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
N028625ML 25 mL
N0286100ML 100 mL
N0286500ML 500 mL
3 options

Catalog No. N028625ML

Supplier: TCI America N028625ML

Only null left

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -51°C
Color Colorless
Boiling Point 150°C
Quantity 25 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonane
EINECSNumber (2)-0009
RTECSNumber RA6115000
TSCA Yes