missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonane, 99%, Reagent Grade, Honeywell™
Reagent Grade, 99%
$1053.89 - $1053.89
Chemical Identifiers
| CAS | 111-84-2 |
|---|---|
| Molecular Formula | C9H20 |
| Molecular Weight (g/mol) | 128.259 |
| MDL Number | MFCD00009574 |
| InChI Key | BKIMMITUMNQMOS-UHFFFAOYSA-N |
| Synonym | n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard |
| PubChem CID | 8141 |
| ChEBI | CHEBI:32892 |
| IUPAC Name | nonane |
| SMILES | CCCCCCCCC |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 111-84-2 | |
| 128.259 | |
| BKIMMITUMNQMOS-UHFFFAOYSA-N | |
| 8141 | |
| nonane |
| C9H20 | |
| MFCD00009574 | |
| n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
| CHEBI:32892 | |
| CCCCCCCCC |
Specifications
| 111-84-2 | |
| Colorless | |
| 151°C (1013 hPa) | |
| Characteristic | |
| C9H20 | |
| MFCD00009574 | |
| UN1920 | |
| n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
| CCCCCCCCC | |
| 128.259 | |
| CHEBI:32892 | |
| 0.99 | |
| Nonane |
| -53°C | |
| 0.718g/mL at 25°C | |
| 31°C (87.8°F) | |
| Glass bottle | |
| CH3(CH2)7CH3 | |
| 500 mL | |
| 1696917 | |
| BKIMMITUMNQMOS-UHFFFAOYSA-N | |
| nonane | |
| 8141 | |
| 128.26g/mol | |
| Liquid |
Safety and Handling
P210-P273-P301 + P310-P304 + P340 + P312-P331-P391
DOTInformation : 3/PGIII
RTECSNumber : 203-913-4