missing translation for 'onlineSavingsMsg'
Learn More

Nonane, 99%, Reagent Grade, Solstice

Catalog No. 60048102
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048102 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048102 Supplier Solstice Supplier No. N29406500ML
Only null left

Reagent Grade, 99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -53°C
Color Colorless
Density 0.718g/mL at 25°C
Boiling Point 151°C (1013 hPa)
Flash Point 31°C (87.8°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3(CH2)7CH3
Quantity 500 mL
UN Number UN1920
Beilstein 1696917
Formula Weight 128.26g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nonane
Show More Show Less
Hazard Statement P210-P273-P301 + P310-P304 + P340 + P312-P331-P391
DOTInformation 3/PGIII
RTECSNumber 203-913-4