missing translation for 'onlineSavingsMsg'
Learn More

Nonanal 95.0+%, TCI America™

Catalog No. N0296500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0296500ML 500 mL
N029625ML 25 mL
N0296100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0296500ML Supplier TCI America Supplier No. N0296500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-19-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00007030
InChI Key GYHFUZHODSMOHU-UHFFFAOYSA-N
Synonym pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
PubChem CID 31289
ChEBI CHEBI:84268
IUPAC Name nonanal
SMILES CCCCCCCCC=O

Specifications

Color Yellow
Boiling Point 192°C
Quantity 500 mL
Formula Weight 142.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanal
EINECSNumber (2)-0494
RTECSNumber RA5700000
TSCA Yes