missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanal Dimethyl Acetal 97.0+%, TCI America™
$44.73 - $124.89
Chemical Identifiers
| CAS | 18824-63-0 |
|---|---|
| Molecular Formula | C11H24O2 |
| Molecular Weight (g/mol) | 188.311 |
| MDL Number | MFCD00036646 |
| InChI Key | LYLVOCPDQAOQKL-UHFFFAOYSA-N |
| Synonym | 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal, Pelargonaldehyde Dimethyl Acetal |
| PubChem CID | 87813 |
| IUPAC Name | 1,1-dimethoxynonane |
| SMILES | CCCCCCCCC(OC)OC |
Chemical Identifiers
| 18824-63-0 | |
| 188.311 | |
| LYLVOCPDQAOQKL-UHFFFAOYSA-N | |
| 87813 | |
| CCCCCCCCC(OC)OC |
| C11H24O2 | |
| MFCD00036646 | |
| 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal, Pelargonaldehyde Dimethyl Acetal | |
| 1,1-dimethoxynonane |
Specifications
| 18824-63-0 | |
| 88°C | |
| MFCD00036646 | |
| 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal, Pelargonaldehyde Dimethyl Acetal | |
| CCCCCCCCC(OC)OC | |
| 188.311 | |
| 188.31 | |
| Liquid |
| Colorless | |
| C11H24O2 | |
| 5 g | |
| LYLVOCPDQAOQKL-UHFFFAOYSA-N | |
| 1,1-dimethoxynonane | |
| 87813 | |
| ≥97.0% (GC) | |
| Nonanal Dimethyl Acetal |
Safety and Handling
TSCA : Yes