missing translation for 'onlineSavingsMsg'
Learn More

Nonanal, 97%

Catalog No. AAA1590836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590836 500 g
AAA1590822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1590836 Supplier Thermo Scientific Chemicals Supplier No. A1590836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-19-6 | C9H18O | 142.242 g/mol

Nonanal is used for fragranced end-products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanal is used for fragranced end-products.

Solubility
Insoluble in water.

Notes
Incompatible with reducing agents, strong basesand strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS 124-19-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00007030
InChI Key GYHFUZHODSMOHU-UHFFFAOYSA-N
Synonym pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
PubChem CID 31289
ChEBI CHEBI:84268
IUPAC Name nonanal
SMILES CCCCCCCCC=O

Specifications

Density 0.825
Boiling Point 79°C to 81°C (12 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.424
Quantity 500 g
Beilstein 1236701
Solubility Information Insoluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material Nonanal
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 204-688-5
RTECSNumber RA5700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only