missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorovaleric Acid 98.0+%, TCI America™

Catalog No. N06055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06055G 5 g
N060525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06055G Supplier TCI America Supplier No. N06055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2706-90-3
Molecular Formula C5HF9O2
Molecular Weight (g/mol) 264.05
MDL Number MFCD00040211
InChI Key CXZGQIAOTKWCDB-UHFFFAOYSA-N
Synonym perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid
PubChem CID 75921
ChEBI CHEBI:83491
IUPAC Name nonafluoropentanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 70°C
Quantity 5 g
UN Number 3265
Formula Weight 264.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Nonafluorovaleric Acid
EINECSNumber (2)-1182
TSCA Yes