missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonafluorovaleric Acid 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 70°C |
| Quantity | 25 g |
| UN Number | 3265 |
| Formula Weight | 264.05 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Nonafluorovaleric Acid |
Safety and Handling
| EINECSNumber | (2)-1182 |
| TSCA | Yes |