missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorovaleric Acid (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS], TCI America™

Catalog No. A571410ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A571410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A571410ML Supplier TCI America Supplier No. A571410ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2706-90-3
Molecular Formula C5HF9O2
Molecular Weight (g/mol) 264.05
MDL Number MFCD00040211
InChI Key CXZGQIAOTKWCDB-UHFFFAOYSA-N
Synonym perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid
PubChem CID 75921
ChEBI CHEBI:83491
IUPAC Name nonafluoropentanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Quantity 10 mL
UN Number 3265
Formula Weight 264.05
Physical Form Liquid
Chemical Name or Material Nonafluorovaleric Acid (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS]
EINECSNumber (2)-1182
TSCA Yes