Learn More
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
$157.09 - $157.09
Chemical Identifiers
| CAS | 2706-90-3 |
|---|---|
| Molecular Formula | C5HF9O2 |
| Molecular Weight (g/mol) | 264.05 |
| MDL Number | MFCD00040211 |
| InChI Key | CXZGQIAOTKWCDB-UHFFFAOYSA-N |
| Synonym | perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid |
| PubChem CID | 75921 |
| ChEBI | CHEBI:83491 |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid |
| SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC309290050
|
Thermo Scientific Chemicals
309290050 |
5 mL | Glass bottle |
Each for $157.09
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2706-90-3 | |
| 264.05 | |
| CXZGQIAOTKWCDB-UHFFFAOYSA-N | |
| 75921 | |
| 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid |
| C5HF9O2 | |
| MFCD00040211 | |
| perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid | |
| CHEBI:83491 | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Specifications
| 2706-90-3 | |
| 100.0 | |
| 1.7100g/mL | |
| Authentic | |
| Glass bottle | |
| 1.2940 to 1.297 | |
| MFCD00040211 | |
| 1.71 | |
| CXZGQIAOTKWCDB-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid | |
| 75921 | |
| 264.05 | |
| Liquid |
| 96.0 | |
| Yellow to Brown | |
| 140°C | |
| 97% | |
| C5HF9O2 | |
| CF3(CF2)3CO2H | |
| 5 mL | |
| perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
| 264.05 | |
| CHEBI:83491 | |
| 97% | |
| Nonafluoropentanoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 220-300-7
RUO â Research Use Only