missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorobutanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH2743703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2743703 1 g
AAH2743706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2743703 Supplier Thermo Scientific Chemicals Supplier No. H2743703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2991-84-6
Molecular Formula C4ClF9O2S
Molecular Weight (g/mol) 318.54
MDL Number MFCD03412262
InChI Key IRFCLLARAUQTNK-UHFFFAOYSA-N
Synonym nonafluoro-1-butanesulfonyl chloride, nonafluorobutane-1-sulfonyl chloride, perfluorobutanesulfonyl chloride, perfluorobutanesulphonyl chloride, nonafluorobutanesulfonyl chloride, 1-butanesulfonylchloride, 1,1,2,2,3,3,4,4,4-nonafluoro, acmc-20ak1o, perfluorobutylsulfonyl chloride, nonafluoro-n-butanesulfonyl chloride
PubChem CID 324955
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl chloride
Formula Weight 318.55
Percent Purity 98%
Chemical Name or Material Nonafluorobutanesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only