missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonafluoro-tert-butyl Alcohol 98.0+%, TCI America™
$115.57 - $917.11
Chemical Identifiers
| CAS | 2378-02-1 |
|---|---|
| Molecular Formula | C4HF9O |
| Molecular Weight (g/mol) | 236.04 |
| MDL Number | MFCD00042092 |
| InChI Key | XZNOAVNRSFURIR-UHFFFAOYSA-N |
| Synonym | perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech |
| PubChem CID | 16924 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol |
| SMILES | C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O |
Chemical Identifiers
| 2378-02-1 | |
| 236.04 | |
| XZNOAVNRSFURIR-UHFFFAOYSA-N | |
| 16924 | |
| C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O |
| C4HF9O | |
| MFCD00042092 | |
| perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech | |
| 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol |
Specifications
| 2378-02-1 | |
| 45°C | |
| MFCD00042092 | |
| perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech | |
| C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O | |
| 236.04 | |
| 236.04 | |
| Liquid |
| Colorless | |
| C4HF9O | |
| 1 g | |
| XZNOAVNRSFURIR-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol | |
| 16924 | |
| ≥98.0% (GC) | |
| Nonafluoro-tert-butyl Alcohol |
Safety and Handling
RTECSNumber : UB6452700
TSCA : No